missing translation for 'onlineSavingsMsg'
Learn More

AS8351, MedChemExpress

Product Code. 16455887
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16455887 5 mg 5mg
16439706 50 mg 50mg
16434446 100 mg 100mg
16403158 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16455887 Supplier Medchem Express Supplier No. HY100744

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AS8351 (NSC51355) is a KDM5B inhibitor, which can induce and sustain active chromatin marks to facilitate the induction of cardiomyocyte-like cells.

AS8351 (NSC51355) is a KDM5B inhibitor, which can induce and sustain active chromatin marks to facilitate the induction of cardiomyocyte-like cells.

Specifications

Chemical Name or Material AS8351
CAS 796-42-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H13N3O2
Quantity 5 mg
Synonym NSC51355
Solubility Information DMSO : 150 mg/mL (514.93 mM; Need ultrasonic and warming)
SMILES O=C(C1=CC=NC=C1)N/N=C/C2=C3C=CC=CC3=CC=C2O
Molecular Weight (g/mol) 291.3
Formula Weight 291.3
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.