missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AS601245, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
AS601245 is an orally active, selective, ATP competitive JNK (c-Jun NH2-terminal protein kinase) inhibitor with IC50s of 150, 220, and 70 nM for three JNK human isoforms (hJNK1, hJNK2, and hJNK3), respectively. AS601245 exhibits 10- to 20-fold selectivity over c-src, CDK2, and c-Raf and more than 50- to 100-fold selectivity over a range of Ser/Thr- and Tyr-protein kinases. Neuroprotective properties.
Specifications
Specifications
| Chemical Name or Material | AS601245 |
| Physical Form | Solid |
| CAS | 345987-15-7 |
| Color | Yellow |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C20H16N6S |
| For Use With (Application) | Cancer-Kinase/protease |
| Solubility Information | DMSO : 10 mg/mL (26.85 mM; Need ultrasonic) |
| SMILES | N#CC(C1=CC=NC(NCCC2=CC=CN=C2)=N1)C3=NC4=CC=CC=C4S3 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction