Learn More
Invitrogen™ AS3MT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143933
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: mouse liver tissue, mouse heart tissue, mouse Neuro-2a whole cell. Flow: mouse spleen tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
AS3MT catalyzes the transfer of a methyl group from S-adenosyl-L-methionine (AdoMet) to trivalent arsenical and may play a role in arsenic metabolism (Lin et al., 2002 [PubMed 11790780]).
Specifications
| AS3MT | |
| Polyclonal | |
| Unconjugated | |
| As3mt | |
| 2310045H08Rik; arsenic (+3 oxidation state) methyltransferase; arsenite methyltransferase; As3mt; Cyt19; Methylarsonite methyltransferase; methyltransferase cyt19; S-adenosyl-L-methionine:arsenic(III) methyltransferase; S-adenosylmethionine:arsenic (III) methyltransferase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57344 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.01mg sodium azide | |
| Q91WU5 | |
| As3mt | |
| E.coli-derived mouse Cyt 19/As3mt recombinant protein (Position: M1-D346). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.