missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AS3MT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106950
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 92%; Zebrafish: 92%.
AS3MT catalyzes the transfer of a methyl group from S-adenosyl-L-methionine (AdoMet) to trivalent arsenical and may play a role in arsenic metabolism (Lin et al., 2002 [PubMed 11790780]).
Specifications
| AS3MT | |
| Polyclonal | |
| Unconjugated | |
| As3mt | |
| 2310045H08Rik; arsenic (+3 oxidation state) methyltransferase; arsenite methyltransferase; As3mt; Cyt19; Methylarsonite methyltransferase; methyltransferase cyt19; S-adenosyl-L-methionine:arsenic(III) methyltransferase; S-adenosylmethionine:arsenic (III) methyltransferase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57412 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q9HBK9 | |
| As3mt | |
| Synthetic peptide directed towards the middle region of human AS3MT (aa 143-193). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction