missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AS3MT Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97425C
This item is not returnable.
View return policy
Description
AS3MT Polyclonal antibody specifically detects AS3MT in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| AS3MT | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| arsenic (+3 oxidation state) methyltransferase, arsenite methyltransferase, CYT19methyltransferase cyt19, EC 2.1.1.137, Methylarsonite methyltransferase, S-adenosyl-L-methionine:arsenic(III) methyltransferase, S-adenosylmethionine:arsenic (III) methyltransferase | |
| Produced in rabbits immunized with E. coli-derived Human AS3MT fragment. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 57412 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction