missing translation for 'onlineSavingsMsg'
Learn More

AS1842856, MedChemExpress

Product Code. 16475827
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475827

Brand: Medchem Express HY100596

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AS1842856, a specific Foxo1 inhibitor (IC50=30 nM), potently suppresses autophagy. AS1842856 only reduces the activity of FoxO1 by binding with it, without affecting its transcription and protein expression.

AS1842856, a specific Foxo1 inhibitor (IC50=30 nM), potently suppresses autophagy. AS1842856 only reduces the activity of FoxO1 by binding with it, without affecting its transcription and protein expression.

Specifications

Chemical Name or Material AS1842856
CAS 836620-48-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H22FN3O3
Quantity 5 mg
Solubility Information DMSO : 5 mg/mL (14.39 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C1=CN(CC)C2=C(C(N)=C(F)C(NC3CCCCC3)=C2)C1=O)O
Molecular Weight (g/mol) 347.38
Formula Weight 347.38
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.