missing translation for 'onlineSavingsMsg'
Learn More

AS1517499, MedChemExpress

Product Code. 16479656
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479656 50 mg 50mg
16435837 5 mg 5mg
16432626 25 mg 25mg
16403108 10 mg 10mg
16474396 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16479656 Supplier Medchem Express Supplier No. HY100614
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AS1517499 is a potent and brain-permeable STAT6 phosphorylation inhibitor with an IC50 of 21 nM.

AS1517499 is a potent and brain-permeable STAT6 phosphorylation inhibitor with an IC50 of 21 nM.

Specifications

Chemical Name or Material AS1517499
CAS 919486-40-1
Color Light Nude
Purity Grade Notes Research
Molecular Formula C20H20ClN5O2
Quantity 50 mg
Solubility Information DMSO : ≥ 35 mg/mL (87.97 mM)
SMILES O=C(C1=CN=C(NCCC2=CC=C(O)C(Cl)=C2)N=C1NCC3=CC=CC=C3)N
Molecular Weight (g/mol) 397.86
Formula Weight 397.86
Percent Purity 98.1%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.