missing translation for 'onlineSavingsMsg'
Learn More

AS-605240, MedChemExpress

Product Code. p-200099969
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16473258

Brand: Medchem Express HY10109

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AS-605240 is a specific and orally active inhibitor of the PI3Kγ, with an IC50 of 8 nM, and a Ki of 7.8 nM.

AS-605240 is a specific and orally active inhibitor of the PI3Kγ, with an IC50 of 8 nM, and a Ki of 7.8 nM.

Specifications

Chemical Name or Material AS-605240
Physical Form Solid
CAS 648450-29-7
Color Pink
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C12H7N3O2S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 5.8 mg/mL (22.54 mM; Need warming) ∣H2O : 2.78 mg/mL (10.81 mM; ultrasonic and warming and adjust pH to 10 with NaOH and heat to 60°C)
SMILES O=C(NC/1=O)SC1=C/C2=CC=C3N=CC=NC3=C2
Molecular Weight (g/mol) 257.27
Formula Weight 257.27
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.