missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AS-605240, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
AS-605240 is a specific and orally active inhibitor of the PI3Kγ, with an IC50 of 8 nM, and a Ki of 7.8 nM.
Specifications
Specifications
| Chemical Name or Material | AS-605240 |
| Physical Form | Solid |
| CAS | 648450-29-7 |
| Color | Pink |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C12H7N3O2S |
| For Use With (Application) | Cancer-Kinase/protease |
| Solubility Information | DMSO : 5.8 mg/mL (22.54 mM; Need warming) ∣H2O : 2.78 mg/mL (10.81 mM; ultrasonic and warming and adjust pH to 10 with NaOH and heat to 60°C) |
| SMILES | O=C(NC/1=O)SC1=C/C2=CC=C3N=CC=NC3=C2 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction