missing translation for 'onlineSavingsMsg'
Learn More

AS-252424, MedChemExpress

Product Code. 16458417
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16458417 5 mg 5mg
16435738 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16458417 Supplier Medchem Express Supplier No. HY13532
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AS-252424 is a potent and selective PI3Kγ inhibitor with an IC50 of 30±10 nM.

AS-252424 is a potent and selective PI3Kγ inhibitor with an IC50 of 30±10 nM.

Specifications

Chemical Name or Material AS-252424
CAS 900515-16-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H8FNO4S
Quantity 5 mg
Solubility Information DMSO : 25 mg/mL (81.89 mM; Need ultrasonic)
SMILES O=C(NC/1=O)SC1=C/C2=CC=C(C3=CC=C(F)C=C3O)O2
Molecular Weight (g/mol) 305.28
Formula Weight 305.28
Percent Purity 99.31%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.