missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AS 1949490

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
AS 1949490 is a selective SHIP2 (SH2 domain-containing inositol 5'-phosphatase 2) inhibitor (IC50 values are 0.34 μM and 0.62 μM for mouse and human respectively); displays approximately 30-fold affinity for SHIP2 over SHIP1. Increases insulin-induced phosphorylation of Akt in L6 myotubules. Stimulates activation of glucose metabolism; regulates gluconeogenesis in vitro and in vivo and exhibits antidiabetic effects. Also promotes increased BDNF mRNA levels in cultured cortical neurons and has memory enhancing and antidepressant effects in vivo.
Specifications
Specifications
| Chemical Name or Material | 3-[(4-Chlorophenyl)methoxy]-N-[(1S)-1-phenylethyl]thiophene-2-carboxamide |
| CAS | 1203680-76-5 |
| Assay Percent Range | >99% |
| Quantity | 25 mg |
| Molecular Formula | C20H18ClNO2S |
| Formula Weight | Observed MW: 371.88 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction