missing translation for 'onlineSavingsMsg'
Learn More

Arylsulfatase G/ARSG Antibody [mFluor Violet 500 SE], Novus Biologicals™

Sheep Polyclonal Antibody

Brand:  Novus Biologicals AF4600MFV500

Product Code. 30647713

  • £482.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Arylsulfatase G/ARSG Polyclonal antibody specifically detects Arylsulfatase G/ARSG in Mouse samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Arylsulfatase G/ARSG
Polyclonal
50mM Sodium Borate
Sheep
Antigen Affinity-purified
RUO
Primary
Mouse
Purified
Immunoprecipitation, Western Blot
mFluor Violet 500 SE
arylsulfatase G, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.4, KIAA1001ASG
Chinese hamster ovary cell line CHO-derived recombinant mouse Arylsulfatase G/ARSG, Gly17-Val525, Accession # Q3TYD4
0.1 mL
Lipid and Metabolism
22901
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.