missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arylsulfatase F Polyclonal Antibody

Arylsulfatase F Polyclonal Antibody
Brand: Invitrogen PA5101436
This item is not returnable.
View return policy
Description
Arylsulfatase F Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| Arylsulfatase F | |
| Polyclonal | |
| Unconjugated | |
| ARSF | |
| ARSF; Arylsulfatase F; ASF | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 416 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P54793 | |
| ARSF | |
| A synthesized peptide derived from human ARSF(Accession P54793), corresponding to amino acid residues G309-L359. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction