missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Arylsulfatase E Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566333
This item is not returnable.
View return policy
Description
Immunogen sequence: FVGALIVHADC FLMRNHTITE QPMCFQRTTP LILQEV Highest antigen sequence identity to the following orthologs - mouse 32%, rat 49%.
Arylsulfatase E is a member of the sulfatase family. It is glycosylated postranslationally and localized to the golgi apparatus. Sulfatases are essential for the correct composition of bone and cartilage matrix. X-linked chondrodysplasia punctata, a disease characterized by abnormalities in cartilage and bone development, has been linked to mutations in this gene.
Specifications
| Arylsulfatase E | |
| Polyclonal | |
| Unconjugated | |
| ARSL | |
| ARSE; Arylsulfatase E; arylsulfatase E (chondrodysplasia punctata 1); ASE; CDPX; CDPX1; CDPXR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 415 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P51690 | |
| ARSL | |
| Recombinant Human Arylsulfatase E. Recombinant protein control fragment (Product #RP-107012). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction