missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Arylsulfatase A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586537
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
The protein encoded by this gene hydrolyzes cerebroside sulfate to cerebroside and sulfate. Defects in this gene lead to metachromatic leucodystrophy, a progressive demyelination disease which results in a variety of neurological symptoms and ultimately death. Alternatively spliced transcript variants have been described for this gene.
Specifications
| Arylsulfatase A | |
| Polyclonal | |
| Unconjugated | |
| ARSA | |
| ARSA; ARSA1; ARSA-I; Arylsulfatase A; Arylsulfatase A component B; Arylsulfatase A component C; As2; As-2; ASA; AS-A; ASNA-I; ATPase ASNA1; AW212749; cerebroside-sulfatase; EC 3.1.6.8; GET3; hASNA-I; MLD; TISP73; TRC40 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 315222, 410 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P15289 | |
| ARSA | |
| Synthetic peptide, corresponding to amino acids 260-310 of Human ARSA. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction