Learn More
Invitrogen™ Arylsulfatase A Monoclonal Antibody (4C10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA533005
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat testis tissue, rat liver tissue, rat brain tissue, rat lung tissue, mouse testis tissue, mouse liver tissue, mouse brain tissue, mouse lung tissue, mouse HEPA1-6 whole cell, human A375 whole cell, human A549 whole cell, human SMMC-7721 whole cell. IHC: human lung cancer tissue. Flow: ANA-1 cell, Raji cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The protein encoded by this gene hydrolyzes cerebroside sulfate to cerebroside and sulfate. Defects in this gene lead to metachromatic leucodystrophy, a progressive demyelination disease which results in a variety of neurological symptoms and ultimately death. Alternatively spliced transcript variants have been described for this gene.
Specifications
| Arylsulfatase A | |
| Monoclonal | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P15289, P50428 | |
| ARSA | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human ARSA (454-482aa QALKQLQLLKAQLDAAVTFGPSQVARGED). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| 4C10 | |
| Unconjugated | |
| ARSA | |
| ARSA; ARSA1; ARSA-I; Arylsulfatase A; Arylsulfatase A component B; Arylsulfatase A component C; As2; As-2; ASA; AS-A; ASNA-I; ATPase ASNA1; AW212749; cerebroside-sulfatase; EC 3.1.6.8; GET3; hASNA-I; MLD; TISP73; TRC40 | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 11883, 315222, 410 | |
| -20°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.