missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARX Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142412
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:1,000.
Arx contains two conserved domains, a C-peptide (or aristaless domain) and the prd-like class homeobox domain. It is a member of the group-II aristaless-related protein family whose members are expressed primarily in the central and/or peripheral nervous system. This protein is thought to be involved in CNS development. Mutations in the Arx gene cause X-linked mental retardation and epilepsy.
Specifications
| ARX | |
| Polyclonal | |
| Unconjugated | |
| ARX | |
| aristaless related homeobox; aristaless-related homeobox; aristaless-related homeobox, X-linked; ARX; arx {ECO:0000250; cancer/testis antigen 121; CT121; EIEE1; Homeobox protein ARX; ISSX; MRX29; MRX32; MRX33; MRX36; MRX38; MRX43; MRX54; MRX76; MRX87; MRXS1; PRTS; RGD1562672; UniProtKB:Q96QS3} | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 170302 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q96QS3 | |
| ARX | |
| Peptide with sequence C-QVSISRSKSYREN (aa 166-178). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction