missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106787
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ARX.
Arx contains two conserved domains, a C-peptide (or aristaless domain) and the prd-like class homeobox domain. It is a member of the group-II aristaless-related protein family whose members are expressed primarily in the central and/or peripheral nervous system. This protein is thought to be involved in CNS development. Mutations in the Arx gene cause X-linked mental retardation and epilepsy.
Specifications
| ARX | |
| Polyclonal | |
| Unconjugated | |
| ARX | |
| aristaless related homeobox; aristaless-related homeobox; aristaless-related homeobox, X-linked; ARX; arx {ECO:0000250; cancer/testis antigen 121; CT121; EIEE1; Homeobox protein ARX; ISSX; MRX29; MRX32; MRX33; MRX36; MRX38; MRX43; MRX54; MRX76; MRX87; MRXS1; PRTS; RGD1562672; UniProtKB:Q96QS3} | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11878, 170302, 317268 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A6YP92, O35085, Q96QS3 | |
| ARX | |
| A synthesized peptide derived from human ARX(Accession Q96QS3), corresponding to amino acid residues A278-Q328. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction