missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARX, Mouse anti-Human, Clone: 1A4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00170302-M31.100ug
This item is not returnable.
View return policy
Description
Sequence: LKISQAPQVSISRSKSYRENGAPFVPPPPALDELGGPGGVTHPEERLGVAGGPGSAPAAGGGTGTEDDEEELLEDEEDEDEEEELLEDDEEELLEDDARALLKEPRSpecifications
| ARX | |
| Monoclonal | |
| Unconjugated | |
| aristaless related homeobox | |
| ISSX/MRX29/MRX32/MRX33/MRX36/MRX38/MRX43/MRX54/MRX76/MRX87/MRXS1/PRTS | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1A4 | |
| In 1x PBS, pH 7.4 | |
| NM_139058 | |
| ARX | |
| ARX (NP_620689, 159 a.a. ∼ 264 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 170302 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction