missing translation for 'onlineSavingsMsg'
Learn More

ARX Antibody [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30733304 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733304 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733304 Supplier Novus Biologicals Supplier No. AF7068MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

ARX Polyclonal antibody specifically detects ARX in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARX
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias aristaless related homeobox, Aristaless-related homeobox, aristaless-related homeobox, X-linked, cancer/testis antigen 121, CT121, EIEE1, homeobox protein ARX, ISSXmental retardation, X-linked 36, mental retardation, X-linked 29, mental retardation, X-linked 32, mental retardation, X-linked 33, mental retardation, X-linked 38, mental retardation, X-linked 43, mental retardation, X-linked 54, mental retardation, X-linked 76, mental retardation, X-linked 87, MRX29, MRX32, MRX33, MRX36, MRX38, MRX43, MRX54, MRX76, MRX87, MRXS1, PRTS
Host Species Sheep
Immunogen E. coli-derived recombinant human ARX, Ser2-Ala100, Accession # Q96QS3
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170302
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.