missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARTS1 Recombinant Rabbit Monoclonal Antibody (23GB1650), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | ARTS1 |
|---|---|
| Clone | 23GB1650 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30087951
|
Invitrogen™
MA553114 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088943
|
Invitrogen™
MA553115 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARTS1 Recombinant Monoclonal antibody specifically detects ARTS1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| ARTS1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q9EQH2, Q9JJ22, Q9NZ08 | |
| 51752, 80897, 80898 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1650 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| adipocyte-derived leucine aminopeptidase; ALAP; A-LAP; aminopeptidase PILS; aminopeptidase regulator of TNFR1 shedding; Appils; Arts1; ARTS-1; CTD-2260A17.2; endoplasmic reticulum aminopeptidase 1; endoplasmic reticulum aminopeptidase 1 delta-Exon-11 isoform; endoplasmic reticulum aminopeptidase 1 delta-Exon-13 isoform; endoplasmic reticulum aminopeptidase 1 delta-Exon-14 isoform; endoplasmic reticulum aminopeptidase 1 delta-Exon-15 isoform; endoplasmic reticulum aminopeptidase associated with antigen processing; ERAAP; ERAAP1; ER-aminopeptidase 1; ERAP1; KIAA0525; leucyl-specific aminopeptidase PILS; PILSA; PILSAP; PILS-AP; Puromycin-insensitive leucyl-specific aminopeptidase; type 1 tumor necrosis factor receptor shedding aminopeptidase regulator; UNQ584/PRO1154; VEGF-induced aminopeptidase | |
| A synthesized peptide derived from human ERAP1 (890-941AA). | |
| Primary | |
| -20°C | |
| Erap1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title