missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARTS1 Polyclonal Antibody

ARTS1 Polyclonal Antibody
Brand: Invitrogen PA5103065
This item is not returnable.
View return policy
Description
ARTS1 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| ARTS1 | |
| Polyclonal | |
| Unconjugated | |
| Erap1 | |
| adipocyte-derived leucine aminopeptidase; ALAP; A-LAP; aminopeptidase PILS; aminopeptidase regulator of TNFR1 shedding; Appils; Arts1; ARTS-1; CTD-2260A17.2; endoplasmic reticulum aminopeptidase 1; endoplasmic reticulum aminopeptidase 1 delta-Exon-11 isoform; endoplasmic reticulum aminopeptidase 1 delta-Exon-13 isoform; endoplasmic reticulum aminopeptidase 1 delta-Exon-14 isoform; endoplasmic reticulum aminopeptidase 1 delta-Exon-15 isoform; endoplasmic reticulum aminopeptidase associated with antigen processing; ERAAP; ERAAP1; ER-aminopeptidase 1; ERAP1; KIAA0525; leucyl-specific aminopeptidase PILS; PILSA; PILSAP; PILS-AP; Puromycin-insensitive leucyl-specific aminopeptidase; type 1 tumor necrosis factor receptor shedding aminopeptidase regulator; UNQ584/PRO1154; VEGF-induced aminopeptidase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51752 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NZ08 | |
| Erap1 | |
| A synthesized peptide derived from human ERAP1(Accession Q9NZ08), corresponding to amino acid residues M447-F497. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction