missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARTS Polyclonal Antibody
GREENER_CHOICE

Product Code. 12858022
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12858022 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12858022 Supplier Invitrogen™ Supplier No. PA520001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

In Western blot applications, this antibody detects a band at ~32kDa. A suggested positive control is human lung tissue lysate. PA5-20001 can be used with blocking peptide PEP-0123.

Apoptosis is related to many diseases and development. Mitochondrial proteins, such as cytochrome c, Apaf-1, and AIF play important role in apoptosis. A novel mitochondrial septin-like protein was identified recently and designated ARTS for apoptosis related protein in TGF-beta signaling pathway. ARTS that is encoded by the human septin H5/PNUTL2/CDCrel2b gene is located to mitochondria and translocates to the nucleus when apoptosis occurs. ARTS is expressed in many tissues. It enhances cell death induced by TGF-beta and, to a lesser extent, by other apoptotic agents, such as TNF-a and Fas ligand.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARTS
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SEPTIN4
Gene Accession No. O43236
Gene Alias 4-Sep; apoptosis-related protein in the TGF-beta signaling pathway; ARTS; Bh5; BRADEION; Bradeion beta; brain protein H5; C17orf47; CE5B3; CE5B3 beta; cell division control-related protein 2; cell division control-related protein 2b; cerebral protein 7; EG3-1RVC; EG3RVC; expression gene 3 in rat visual cortex; expression gene 3-1 in rat visual cortex; H5; hCDCREL-2; hucep-7; MART; M-Septin; OTTMUSG00000001265; peanut-like 2; peanut-like 2 homolog; peanut-like protein 2; PNUTL2; SEP4; Sept4; septin 4; septin H5; Septin4; septin-4; septin-M; Uncharacterized protein C17orf47
Gene Symbols SEPTIN4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids near the amino terminus of human ARTS.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5414
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.