missing translation for 'onlineSavingsMsg'
Learn More

Artificial Seawater, Reagecon
BIOPHARMA

Product Code. p-7159735 Shop All Reagecon Products
Click to view available options
Quantity:
5 L
25 L
Unit Size:
25 L
5 L
This item is not returnable. View return policy

Product Code. 11533222

Brand: Reagecon DSW5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-quality, pure water product, presented in a convenient pack size.

  • Water Reference Standard, Water Reference Material
  • Can be used as a matrix for a variety of solutes or as a cleaning/sanitization solution

Specifications

Packaging HDPE Container
Quantity 5 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.