missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Artemotil, MedChemExpress
Description
Artemotil (β-Arteether) has antimalarial activity for the treatment of chloroquine-resistant Plasmodium falciparum malaria with an IC50 of 1.61 nM. Artemotil also has central nervous system (CNS) neurotoxicity and anorectic toxicity in rats, dogs and monkeys.
Specifications
Specifications
| Chemical Name or Material | Artemotil |
| Physical Form | Solid |
| CAS | 75887-54-6 |
| Color | Off-White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C17H28O5 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | β-Arteether (+)-Arteether Arteether |
| Solubility Information | DMSO : 50 mg/mL (160.05 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?