Learn More
Artemisinin, 98%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J65406.03
| Quantity | 1g |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble to 100mM in DMSO and to 75mM in ethanol
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
Chemical Identifiers
| 63968-64-9 | |
| 282.336 | |
| BLUAFEHZUWYNDE-DKGJTOOQSA-N | |
| 9838675 |
| C15H22O5 | |
| MFCD00081057 | |
| artemisinine, artemisinin, gnf-pf-5341, artemisinin, 18, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.0^ 4,13 .0^ 8,13 hexadecan-10-one, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.04,13.08,13 hexadecan-10-one | |
| CC1CCC2C(C(=O)OC3C24C1CCC(O3)(OO4)C)C |
Specifications
| 63968-64-9 | |
| MFCD00081057 | |
| 14,817 | |
| Soluble to 100mM in DMSO and to 75mM in ethanol | |
| +76° (c=0.5 in Methanol) | |
| 282.336 | |
| 282.33 | |
| Artemisinin |
| C15H22O5 | |
| 1g | |
| artemisinine, artemisinin, gnf-pf-5341, artemisinin, 18, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.0^ 4,13 .0^ 8,13 hexadecan-10-one, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.04,13.08,13 hexadecan-10-one | |
| BLUAFEHZUWYNDE-DKGJTOOQSA-N | |
| CC1CCC2C(C(=O)OC3C24C1CCC(O3)(OO4)C)C | |
| 9838675 | |
| 99% |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.