missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Artemisia vulgaris 1.0101 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£143.00 - £279.00
Specifications
| Antigen | Artemisia vulgaris 1.0101 |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18667997
|
Novus Biologicals
NBP3-12774-100ul |
100 μg |
£279.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600599
|
Novus Biologicals
NBP3-12774-50ul |
50 μg |
£143.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Artemisia vulgaris 1.0101 Polyclonal antibody specifically detects Artemisia vulgaris 1.0101 in Plant samples. It is validated for ELISASpecifications
| Artemisia vulgaris 1.0101 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Plant | |
| Produced in rabbits immunized with purified, recombinant Artemisia vulgaris 1.0101 (Uniprot#: Q84ZX5.1; Met1-His132). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title