missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Artemis Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44276FR
This item is not returnable.
View return policy
Description
Artemis Polyclonal antibody specifically detects Artemis in Mouse, Rat samples. It is validated for Western Blot
Specifications
| Artemis | |
| Polyclonal | |
| Western Blot | |
| ASCID, DCLREC1C, DNA cross-link repair 1C, DNA cross-link repair 1C protein, EC 3.1, FLJ36438, hSNM1C, protein artemis, Protein A-SCID, PSO2 homolog, RS-SCID, severe combined immunodeficiency, type a (Athabascan), SNM1 homolog C, SNM1CS. cerevisiae), SNM1-like protein | |
| The epitope recognized by this antibody maps to a region between residue 575 to 625 of human DNA cross-link repair 1C protein SNM1-like protein using the numbering given in entry Q8K4J0.2 (GeneID 227525). | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, DNA Repair, Non homologous end joining | |
| 64421 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction