missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Artemis Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44276AF532
This item is not returnable.
View return policy
Description
Artemis Polyclonal antibody specifically detects Artemis in Mouse, Rat samples. It is validated for Western Blot
Specifications
| The epitope recognized by this antibody maps to a region between residue 575 to 625 of human DNA cross-link repair 1C protein SNM1-like protein using the numbering given in entry Q8K4J0.2 (GeneID 227525). | |
| 0.1 mL | |
| Primary | |
| Purified |
| Immunogen affinity purified | |
| Cancer, Cell Cycle and Replication, DNA Repair, Non homologous end joining | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction