Learn More
Invitrogen™ Artemin Recombinant Rabbit Monoclonal Antibody (24GB6845)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556972
 View more versions of this product
Product Code. 30275570
  • £126.00 / 20µL
Description
Artemin Recombinant Monoclonal Antibody for Western Blot
The protein encoded by this gene is a member of the glial cell line-derived neurotophic factor (GDNF) family of ligands which are a group of ligands within the TGF-beta superfamily of signaling molecules. GDNFs are unique in having neurotrophic properties and have potential use for gene therapy in neurodegenerative disease. Artemin has been shown in culture to support the survival of a number of peripheral neuron populations and at least one population of dopaminergic CNS neurons. Its role in the PNS and CNS is further substantiated by its expression pattern in the proximity of these neurons. This protein is a ligand for the RET receptor and uses GFR-alpha 3 as a coreceptor. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Artemin
Recombinant Monoclonal
Unconjugated
ARTN
Artemin; ARTN; Enovin; EVN; NBN; neublastin; neurotrophic factor
Rabbit
Affinity chromatography
RUO
11876, 362572, 9048
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB6845
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q5T4W7, Q6AYE8, Q9Z0L2
ARTN
A synthesized peptide derived from human Artemin (aa 189-202).
20 μL
Primary
Human, Mouse, Rat