missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Artemin Antibody (RM0074-6G32), Alexa Fluor™ 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-21701AF350
This item is not returnable.
View return policy
Description
Artemin Monoclonal specifically detects Artemin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Artemin | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ARTN | |
| Mouse recombinant Artemin | |
| 0.1 mL | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant Artemin. | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| RM0074-6G32 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
| artemin, ENOVIN, EVNneurotrophic factor, NBN, Neublastin | |
| Rat | |
| Protein A or G purified | |
| Cancer, Growth and Development, Neuronal Cell Markers, Neuroscience | |
| 9048.0 | |
| Mouse | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction