missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ART5 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA596593

Product Code. 16549431

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: VPILPLGLAP DTFDDTYVGC AEEMEEKAAP LLKEEMAHHA LLRESWEAAQ ETWEDKRRGL TLPPGFKAQN GIAIMVYTNS SNTLYWELNQ AVRTGGGSRE LYMRHFPFKA LHFYLIRALQ LLRGSGGCSR GPGEVVFRGV GSLRFEPKRL GDSVRLGQFA SSSLDKAVAH RFGNATLF; Positive Samples: Mouse skeletal muscle, Rat skeletal muscle, Rat brain.

The protein encoded by this gene belongs to the ARG-specific ADP-ribosyltransferase family. Proteins in this family regulate the function of target proteins by attaching ADP-ribose to specific amino acid residues in their target proteins. The mouse homolog lacks a glycosylphosphatidylinositol-anchor signal sequence and is predicted to be a secretory enzyme. Transcript variants with different 5' UTRs, but encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ART5
Polyclonal
Unconjugated
ART5
ADP-ribosyltransferase 5; ADP-ribosyltransferase C2 and C3 toxin-like 5; Art5; ARTC5; Ecto-ADP-ribosyltransferase 5; mono(ADP-ribosyl)transferase 5; NAD(P)(+)--arginine ADP-ribosyltransferase 5; UNQ575/PRO1137; YAC-2
Rabbit
Affinity Chromatography
RUO
116969, 11875, 259167
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P70352, Q5XHY4, Q96L15
ART5
Recombinant protein (or fragment).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.