missing translation for 'onlineSavingsMsg'
Learn More

ART4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18323714 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18323714 20 μL 20µL
18348114 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18323714 Supplier Novus Biologicals Supplier No. NBP30388820UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ART4 Polyclonal specifically detects ART4 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ART4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias ADP-ribosyltransferase 4 (DO blood group), ADP-ribosyltransferase 4 (Dombrock blood group), CD297, CD297 antigen, DODombrock blood group, DOK1ADP-ribosyltransferase 4, Dombrock blood group carrier molecule, EC 2.4.2.31, ecto-ADP-ribosyltransferase 4, Mono(ADP-ribosyl)transferase 4, mono-ADP-ribosyltransferase 4, NAD(P)(+)--arginine ADP-ribosyltransferase 4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human ART4 (NP_066549.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 420
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.