missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ART3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15930-1-AP-150UL
This item is not returnable.
View return policy
Description
Mono-ADP-ribosylation involves the transfer of the ADP-ribose moiety from NAD+ to a specific amino acid in the target protein. The rodent mono-ADP-ribosyltransferase RT6 is a glycosylphosphatidylinositol (GPI)-anchored membrane protein specifically expressed at the cell surface of rat and mouse T lymphocytes. The predicted 367-amino acid human ART3 protein has an estimated molecular mass of 41.5 kD and contains a hydrophobic peptide signal at its N terminus, 3 consensus motifs specific to enzymes catalyzing ADP-ribose transfer, a hydrophobic C-terminal sequence characteristic of a GPI-anchored protein, a novel motif repeated 3 times at its C terminus, and 1 potential glycosylation site.1 The ART3 and rodent RT6 proteins share 35% amino acid identity.Specifications
| ART3 | |
| Polyclonal | |
| Unconjugated | |
| ART3 | |
| ADP ribosyltransferase 3, ART3, Ecto ADP ribosyltransferase 3, Mono(ADP ribosyl)transferase 3, TMART | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 109979, 419 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13508, Q8R2G4 | |
| ART3 | |
| ART3 Fusion Protein Ag8775 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction