missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ART1 Antibody (OTI6E2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46545
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
ART1 Monoclonal specifically detects ART1 in Human samples. It is validated for Western Blot.
Specifications
| ART1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000 | |
| ADP-ribosyltransferase 1, ADP-ribosyltransferase 2, CD296, GPI-linked NAD(P)(+)--arginine ADP-ribosyltransferase 1, MGC133217, mono(ADP-ribosyl)transferase 1, RT6 | |
| Mouse | |
| 36.2 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| OTI6E2 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| ART1 | |
| Human recombinant protein fragment corresponding to amino acids 83-295 of human ART1 (NP_004305) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| 417 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction