missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ART1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99051-100ul
This item is not returnable.
View return policy
Description
ART1 Polyclonal antibody specifically detects ART1 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ART1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| ADP-ribosyltransferase 1, ADP-ribosyltransferase 2, CD296, GPI-linked NAD(P)(+)--arginine ADP-ribosyltransferase 1, MGC133217, mono(ADP-ribosyl)transferase 1, RT6 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the mouse ART1. | |
| 100 μg | |
| CD Markers, Immunology | |
| 417 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction