missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ART™ Reload System
GREENER_CHOICE

Product Code. 15173626
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
200 μL
300 μL
1000 μL
For Use With (Equipment):
Eppendorf
Universal Fit
Model:
ART Non-filtered
ART Ultra Micro Non-Filtered
Fit for Purpose:
Certified Quality
Sterile
Unit Size:
Pack of 560
Pack of 960
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) For Use With (Equipment) Model Fit for Purpose unitSize
15173626 1000 μL Universal Fit ART Non-filtered Certified Quality Pack of 560
15103636 1000 μL Universal Fit ART Non-filtered Sterile Pack of 560
15193626 1000 μL Universal Fit ART Non-filtered Sterile Pack of 560
15183626 1000 μL Universal Fit ART Non-filtered Certified Quality Pack of 560
15183566 10 μL Universal Fit ART Non-filtered Sterile Pack of 960
15193566 10 μL Universal Fit ART Non-filtered Sterile Pack of 960
15123576 10 μL Universal Fit ART Non-filtered Certified Quality Pack of 960
15113576 10 μL Universal Fit ART Ultra Micro Non-Filtered Certified Quality Pack of 960
15173586 20 μL Eppendorf ART Non-filtered Sterile Pack of 960
15143586 20 μL Eppendorf ART Non-filtered Sterile Pack of 960
15153586 20 μL Eppendorf ART Non-filtered Certified Quality Pack of 960
15123586 20 μL Eppendorf ART Non-filtered Certified Quality Pack of 960
15103606 200 μL Universal Fit ART Non-filtered Sterile Pack of 960
15163596 200 μL Universal Fit ART Non-filtered Sterile Pack of 960
15193596 200 μL Universal Fit ART Non-filtered Certified Quality Pack of 960
15183596 200 μL Universal Fit ART Non-filtered Certified Quality Pack of 960
15183616 300 μL Universal Fit ART Non-filtered Sterile Pack of 960
15163616 300 μL Universal Fit ART Non-filtered Sterile Pack of 960
15173616 300 μL Universal Fit ART Non-filtered Certified Quality Pack of 960
15153616 300 μL Universal Fit ART Non-filtered Certified Quality Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
This item is not returnable. View return policy
Product Code. 15173626 Supplier Thermo Scientific™ Supplier No. 3102RT
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Reduce waste and save benchtop and storage space with this advanced nonfiltered reload system.

Reduce waste and save benchtop and storage space with the advanced Thermo Scientific™ ART™ Nonfiltered Reload System. This system provides an environmentally responsible option without compromising sterility.

Features:

  • Stackable barrier tip packaging allows for tips to pass through offset holes in lower insert
  • Blister sleeve guide rails align tip inserts for smooth transfer into the rack
  • Outer sleeve protects tips from exterior contamination while refilling inserts
  • Tip packaging option reduces plastic waste
  • Rigid insert design provides reliable tip loading
  • Lot number and description are printed on every insert

Note:

Empty rack required for -RT parts

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Model ART Non-filtered
Length (English) 3.5 in.
Length (Metric) 8.89 cm
Clean Claims DNase, RNase, DNA, ATP and Endotoxin Free
Format Rack
Quantity 560 Pk.
For Use With (Equipment) Universal Fit
Product Line ART
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.