Learn More
Thermo Scientific™ ART™ Non-Filtered Multichannel HydroLogix HLT Pipette Tips
A variety of tip styles and configurations
Brand: Thermo Scientific™ 3950-11
Code : 03
Additional Details : Weight : 0.92700kg
Description
A variety of tip styles and configurations
The HydroLogix™ (HLT) design utilizes a unique bi-fabricated wall that has been designed to produce a secure seal at a fraction of the force required to load standard pipet tips onto the pipettor. Accordingly, the forces required to eject the tips from your pipettor are significantly reduced.
- Available in blue or clear color
Specifications
1000 μL | |
Non-sterile | |
Bulk |
Non-Filtered | |
Blue | |
ART |