missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ART™ Barrier Pipette Tips in Bulk Packaging

Product Code. p-7146707
Click to view available options
Volume (Metric):
100 μL
200 μL
5 mL
Unit Size:
Pack of 1000
Pack of 250
Pack of 5000
This item is not returnable. View return policy

Product Code. 15201619

Brand: Thermo Scientific™ 2065EB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermo Scientific™ ART™ Barrier Pipette tips seal in contaminants, providing bi-directional protection for samples and pipettes. Fits most pipettes.

ART™ Barrier tips are not the same as traditional filter pipette tips. ART Barrier tips contain a one-of-a-kind, self-sealing barrier that seals in aerosol and liquid contaminants on contact, providing bi-directional protection from contamination for pipettes and samples.

  • Universal Tip, fits most brands of pipettes
  • ART™ tips are certified to be free DNase, RNase, DNA, ATP, Endotoxin, and Pyrogens
  • Convenient bulk packaging for high volume usage

  • Sterile
  • Low Retention
  • Racked with lift-off lid
TRUSTED_SUSTAINABILITY

Specifications

Description ART 100E, Filtered, Non-sterile, Bulk
Volume (Metric) 100 μL
Filtered Filtered
Sterility Non-sterile
Color Clear
Rack Color no rack - bulk packaging
Autoclavable Not Autoclavable
Model ART 100E
Length (English) 2 inches
Length (Metric) 5.07 cm
Clean Claims DNase, RNase, DNA, ATP, Endotoxin, and Pyrogen Free
Format Bulk
Tip Style Micro Point
Packaging 5 bags of 1000 tips each
Quantity 5000 tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Product Line ART
Material Virgin Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.