missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARSH Polyclonal Antibody

ARSH Polyclonal Antibody
Brand: Invitrogen PA569329
This item is not returnable.
View return policy
Description
ARSH Polyclonal Antibody for Western Blot
Specifications
| ARSH | |
| Polyclonal | |
| Unconjugated | |
| ARSH | |
| ARSH; arylsulfatase family member H; arylsulfatase family, member H; arylsulfatase H; arylsulfatase H; arylsulfatase family member H; ASH; sulfatase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 347527 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5FYA8 | |
| ARSH | |
| synthetic peptide directed towards the middle region of human ARSH. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction