missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARSG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12328-1-AP-150UL
This item is not returnable.
View return policy
Description
Sulfatases, such as ARSG, hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules. This gene, a novel arylsulfatase, has activity toward pseudosubstrates including p-nitrocatechol sulfate and 4-methylumbelliferyl sulfate. Activity is competitively inhibited by phosphate. The unprocessed protein is active as a 63-kDa monomer and demonstrates an acidic pH optimum as typically seen for lysosomal sulfatases. The protein accumulates within lysosomes and is also a glycoprotein that binds specifically to mannose 6-phosphate receptors.Specifications
| ARSG | |
| Polyclonal | |
| Unconjugated | |
| ARSG | |
| ARSG, arylsulfatase G, ASG, KIAA1001 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22901, 303631, 74008 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q32KJ9, Q3TYD4, Q96EG1 | |
| Arsg | |
| ARSG Fusion Protein Ag2991 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction