missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arsenic standard solution, BAKER INSTRA-ANALYZED reagent for AAS, J.T. Baker™
Shop All J.T.Baker ProductsSpecifications
Specifications
| Concentration | 1000 mg/L |
| Certifications/Compliance | Traceable to NIST. |
| Solvent or Matrix | 2-5% HNO3 + (CHOH)2(COOH)2 |
| Quantity | 100 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?