missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arsenic Standard For ICP, 10,000μg/mL in 5% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
£113.00 - £217.00
Chemical Identifiers
CAS | 7440-38-2, 7697-37-2, 7732-18-5 |
---|---|
Molecular Formula | As |
Molecular Weight (g/mol) | 74.92 |
MDL Number | MFCD00085309 |
InChI Key | RQNWIZPPADIBDY-UHFFFAOYSA-N |
PubChem CID | 5359596 |
ChEBI | CHEBI:27563 |
IUPAC Name | arsenic |
SMILES | [As] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10426922
|
SPEX Certiprep
PLAS2-3Y |
125 mL | Triple-leached LDPE bottle |
£113.00
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10406752
|
SPEX Certiprep
PLAS2-3X |
500 mL | Triple-leached LDPE bottle |
£217.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
As | 10,000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7440-38-2, 7697-37-2, 7732-18-5 | |
74.92 | |
RQNWIZPPADIBDY-UHFFFAOYSA-N | |
CHEBI:27563 | |
[As] |
As | |
MFCD00085309 | |
5359596 | |
arsenic |
Specifications
7440-38-2, 7697-37-2, 7732-18-5 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00085309 | |
[As] | |
74.92 | |
CHEBI:27563 | |
5% HNO3 |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
As | |
RQNWIZPPADIBDY-UHFFFAOYSA-N | |
arsenic | |
5359596 | |
Assurance | |
Arsenic Standard |
Safety and Handling
Causes skin irritation, serious eye irritation. May cause respiratory irritation
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Carcinogenicity Category 1A
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H350-May cause cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitric acid, arsenic
Product intended for laboratory use only