missing translation for 'onlineSavingsMsg'
Learn More

Arsenic polycrystalline lump, 2-8mm (0.08-0.3in), Puratronic™, 99.99999+% (metals basis)

Product Code. 11362128
Click to view available options
Quantity:
1 g
10 g
25 g
100 g
1 kg
Unit Size:
1000g
100g
10g
1g
25g
This item is not returnable. View return policy

Product Code. 11362128

Brand: Thermo Scientific Alfa Aesar 013044.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-38-2 | As | 74.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-38-2
Molecular Formula As
Molecular Weight (g/mol) 74.92
MDL Number MFCD00085309
InChI Key RQNWIZPPADIBDY-UHFFFAOYSA-N
Synonym arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
PubChem CID 5359596
ChEBI CHEBI:27563
IUPAC Name arsenic
SMILES [As]

Specifications

Color Gray
Quantity 25 g
Odor Odorless
Sensitivity Air Sensitive
UN Number UN1558
Formula Weight 74.92
Percent Purity ≥99.99999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Arsenic polycrystalline lump, 2 to 8mm (0.08 to 0.3 in.)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.