missing translation for 'onlineSavingsMsg'
Learn More

Arsenic polycrystalline lump, 2-8mm (0.08-0.3in), Puratronic™, 99.99999+% (metals basis), Thermo Scientific Chemicals

Catalog Number 11372128
Click to view available options
Quantity:
1 g
10 g
25 g
100 g
1 kg
This item is not returnable. View return policy
7440-38-2
As
74.92
MFCD00085309
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
5359596
CHEBI:27563
arsenic
[As]
This item is not returnable. View return policy
7440-38-2
As
74.92
MFCD00085309
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
5359596
CHEBI:27563
arsenic
[As]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Color Gray
UN Number UN1558
Quantity 1 g
Formula Weight 74.92
Percent Purity ≥99.99999%
Grade Puratronic™
Odor Odorless
Sensitivity Air Sensitive
Assay (metals basis)
Chemical Name or Material Arsenic polycrystalline lump, 2 to 8mm (0.08 to 0.3 in.)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'