missing translation for 'onlineSavingsMsg'
Learn More

Arsenic lump, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11386577
Click to view available options
Quantity:
25 g
100 g
7440-38-2
As
74.92
MFCD00085309
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
5359596
CHEBI:27563
arsenic
[As]
7440-38-2
As
74.92
MFCD00085309
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
5359596
CHEBI:27563
arsenic
[As]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 99.99% (metals basis)
Quantity 25 g
Chemical Name or Material Arsenic lump
compliance-icons compliance-icons
Product Identifier
  • Arsenic lump
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'