missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) selenide, 99.999% (metals basis)

Product Code. 11350368
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11350368 5 g 5g
11340368 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11350368 Supplier Thermo Scientific Alfa Aesar Supplier No. 043748.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Used as a vacuum deposition, chalcogenide glass for infrared optics, as it transmits light with wavelengths between 870 nm and 17.2 μm. | CAS: 1303-36-2 | As2Se3 | 386.76 g/mol

Arsenicselenide is used as a vacuum deposition, chalcogenide glass for infrared optics, as it transmits light with wavelengths between 870 nm and 17.2 μm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-36-2
Molecular Formula As2Se3
Molecular Weight (g/mol) 386.76
MDL Number MFCD00014166
InChI Key WBFMCDAQUDITAS-UHFFFAOYSA-N
Synonym arsenic triselenide, diarsenic triselenide, arsenic selenide, arsenous selenide, arsenic sesquiselenide, unii-xws0w0gb8g, diarsenic triselenide asse3, arsenic iii selenide, xws0w0gb8g
PubChem CID 14772
IUPAC Name [(selanylidenearsanyl)selanyl]arsaneselone
SMILES [Se]=[As][Se][As]=[Se]

Specifications

Melting Point 260°C
Physical Form Powder
Quantity 5 g
UN Number UN3077
Solubility Information Soluble in nitric acid. Insoluble in water.
Formula Weight 386.72
Percent Purity 99.999%
Packaging Glass bottle
Specific Gravity 4.75
Density 4.75
Chemical Name or Material Arsenic(III) selenide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.