missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog Number 11335817
Click to view available options
Quantity:
25 g
250 g
1 kg
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
1327-53-3
As2O3
197.84
MFCD00003433
IKWTVSLWAPBBKU-UHFFFAOYSA-N
Arsenic trioxide
diarsorosooxidane
O=[As]O[As]=O
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
1327-53-3
As2O3
197.84
MFCD00003433
IKWTVSLWAPBBKU-UHFFFAOYSA-N
Arsenic trioxide
diarsorosooxidane
O=[As]O[As]=O

Used in the manufacturing of such products as glass and enamels. Arsenic trioxide is utilized in various studies on cellular processes, notably in investigations of leukemia and principal material for the production of many arsenic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 465°C (sublimation)
Melting Point 312.3°C
Physical Form Powder
Quantity 25 g
UN Number UN1561
Merck Index 14,804
Solubility Information Slightly soluble in cold water; Soluble in boiling water,HCl,alkali hydroxide; Insoluble in alcohol,chloroform,and etherSoluble in dilute acids and alkalies; Insoluble in organic solvents; Slightly soluble in water.
Formula Weight 197.84
Percent Purity 99.5%
Odor Odorless
Density 3.74 g/mL
Assay (metals basis)
Chemical Name or Material Arsenic (III) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'