missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) iodide, 99.999% (metals basis)

Product Code. p-7051783
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11324957

Brand: Thermo Scientific Alfa Aesar 041962.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 7784-45-4 | AsI3 | 455.635 g/mol

Arsenic(III) iodide is useful for preparing organoarsenic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-45-4
Molecular Formula AsI3
Molecular Weight (g/mol) 455.635
MDL Number MFCD00014170
InChI Key IKIBSPLDJGAHPX-UHFFFAOYSA-N
Synonym arsenous triiodide, arsenic iii iodide, arsenic iodide, arsenic triiodide, triiodoarsine, arsenous iodide, hsdb 420, arsen iii-jodid
PubChem CID 24575
IUPAC Name triiodoarsane
SMILES [As](I)(I)I

Specifications

Boiling Point 424°C
Melting Point 140°C
Physical Form Powder
Quantity 5 g
UN Number UN1557
Merck Index 14,803
Solubility Information Hydrolyzes slowly in water.
Formula Weight 455.64
Percent Purity 99.999%
Odor Pungent
Sensitivity Moisture sensitive
Density 4.39 g/mL
Assay (metals basis)
Chemical Name or Material Arsenic(III) iodide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.