missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Arsenic(III) iodide, 99.999% (metals basis)

Brand:  Alfa Aesar™ 041962.14

Code : NEW

Additional Details : CAS Number : 7784-45-4 Weight : 0.02500kg Transport : UN number : 1557 Chem class : 1 Pack group : II

Quantity 25g

 View more versions of this product

Product Code. 11334957

  • £288.00 / 25g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Arsenic(III) iodide
455.64
UN1557
7784-45-4
MFCD00014170
14,803

Specifications

Arsenic(III) iodide
Powder
(metals basis)
MFCD00014170
14,803
−80 Mesh
Moisture Sensitive
140°C
7784-45-4
99.999%
455.64
UN1557
4.39g/mL
Pungent
424°C
25g
Videos
SDS